• USA Home
  • U5007 - cDNA Probe Ubiquitin human

EMAIL THIS PAGE TO A FRIEND
U5007 Sigma

cDNA Probe Ubiquitin human

Purchase

Description

Application

Genes that are highly conserved and expressed in a wide variety of tissue types are useful as reference "house-keeping" genes. They allow determination of relative expression levels of other genes of interest.

Other Notes

The human ubiquitin cDNA probe is a 2.1-kb fragment containing the complete coding sequence of human poly(ubiquitin). The transcript sizes detected with this probe are 2.3 kb (human) and 1.35 kb (mouse). Minor bands are also observed at 6 kb and 0.6 kb.
GenBank® Accession Number M26880.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

required but not provided

Product #

Description

Add to Cart

W4502 Water, Molecular Biology Reagent

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?