• USA Home
  • U5254 - n-Undecyl β-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
U5254 Sigma

n-Undecyl β-D-glucopyranoside

~97% (GC)

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents N to Z, Non-Ionic Detergents
description   non-ionic
InChI Key   ULDAPNVYSDTSFM-UHFFFAOYSA-N
assay   ~97% (GC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?