• USA Home
  • U7253 - Urotensin I teleost fish

EMAIL THIS PAGE TO A FRIEND
U7253 Sigma

Urotensin I teleost fish

≥95% (HPLC)

  • CAS Number 83930-33-0

  • Empirical Formula (Hill Notation) C210H340N62O67S2

  • Molecular Weight 4869.45

  •  MDL number MFCD00082128

Purchase

Description

Amino Acid Sequence

Asn-Asp-Asp-Pro-Pro-Ile-Ser-Ile-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Asn-Met-Ile-Glu-Met-Ala-Arg-Ile-Glu-Asn-Glu-Arg-Glu-Gln-Ala-Gly-Leu-Asn-Arg-Lys-Tyr-Leu-Asp-Glu-Val-NH2

Application

Urotensin I teleost fish was administered intracerebroventricularly in juvenile rainbow trout (Oncorhynchus mykiss) for studying the impact of corticotropin-releasing factor (CRF) and urotensin I on aggression and anxiety-like behaviour.

Biochem/physiol Actions

A neuroendocrine peptide of the CRF family that is found in the brain of most fish species. It may be involved in the regulation of blood pressure in fish. It is closely related to mammalian urocortin and releases ACTH from cultured rat pituitary cells.

Urotensin I is a CRF-like neuropeptide, expresses mainly in the brain of most fish species. It induces the acetylcholine (ACTH) release from the superfused goldfish anterior pituitary cell column. It stimulates the marked, dose-dependent reduction in perfusion pressure in the hind limb of the rat.

Price and Availability


Amplified Detection

Free Webinar

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?