• USA Home
  • U7383 - Anti-Ubiquilin-1 (N-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
U7383 Sigma

Anti-Ubiquilin-1 (N-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-DA41, Anti-DSK2, Anti-PLIC-1, Anti-Protein-linking IAP to cytoskeleton 1, Anti-UBQLN1, Anti-XDRP1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Alzheimer′s Disease, Antibodies for Cell Biology, Antibodies for Neurobiology,
species reactivity   human, mouse, rat
application(s)   western blot: 0.5-1 μg/mL using lysates of HEK293-T cells and COS7 cells expressing human ubiquilin-1
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~62 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... UBQLN1(29979)
mouse ... Ubqln1(56085)
rat ... Ubqln1(114590)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 1-19 of human ubiquilin-1 conjugated to KLH. This sequence is highly conseved (2 amino acid substitutions) in rat and mouse ubiquilin-1.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

HPA006431 Anti-UBQLN2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
U7258 Anti-Ubiquilin-1 antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?