EMAIL THIS PAGE TO A FRIEND
U7507 Sigma

Urotensin II rat

≥97% (HPLC), synthetic

  • Empirical Formula (Hill Notation) C77H102N18O20S2

  • Molecular Weight 1663.87

  •  PubChem Substance ID 329827648

Purchase

Properties

Related Categories CRF, Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins,
biological source   synthetic
InChI Key   BOBVDMGRLAZVCI-BGYIBPDASA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   rat ... Uts2(29180)

Description

Amino Acid Sequence

Glp-His-Gly-Thr-Ala-Pro-Glu-Cys-Phe-Trp-Lys-Tyr-Cys-Ile [Disulfide Bridge: 8-13]

Other Notes

Lyophilized from 0.1% TFA in H2O

Application

Rat Urotensin II was used for testing the specificity of immunoreaction for studying the distribution of corticotropin-releasing hormone in the brain of the snake Bothrops jararaca.

Biochem/physiol Actions

A neuroendocrine peptide of the CRF family that was originally isolated from fish brain but is also found in mammalian brain and spinal motor neurons. It is a potent vasoconstrictor. It may be a mediator of stress-induced anorexia.

Rat Urotensin II is a neuroendocrine peptide that belongs to CRF family and possesses regionally-selective vasodilator impact on conscious rats. It also induces potent vasoconstriction in humans in vivo.

Price and Availability


Amplified Detection

Free Webinar

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?