EMAIL THIS PAGE TO A FRIEND
U7632 Sigma

Undecaprenol

~98%, methylene chloride solution

Synonym: Prenol C55

Purchase

Properties

assay   ~98%
form   methylene chloride solution

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1846 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?