• USA Home
  • U8502 - Methyl 10-undecenoate

EMAIL THIS PAGE TO A FRIEND
U8502 Sigma

Methyl 10-undecenoate

~99% (TLC)

Synonym: 10-Undecenoic acid methyl ester, Undecylenic acid methyl ester

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   KISVAASFGZJBCY-UHFFFAOYSA-N
assay   ~99% (TLC)
refractive index   n20/D 1.437(lit.)
bp   245 °C(lit.)
density   0.882 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
Flash Point(F) 
239 °F
Flash Point(C) 
115 °C
Protocols & Articles

Articles

Molecular Monolayers on Silicon Surfaces

Controlled formation of organic molecular monolayers on silicon surfaces offers the promise of enhancing the functionality of existing and emerging silicon based materials and devices. The synthetic ...
Gregory P. Lopinski, Daniel D.M. Wayner
Material Matters 2008, 3.2, 38.
Keywords: Adsorption, Applications, Buffers, Degradations, Diffusion, Electronics, Events, Gas chromatography, Grignard Reaction, Hydrosilylations, Immobilization, Infrared spectroscopy, Methods, Microelectronics, Microscopy, Nanomaterials, Nucleic acid hybridization, Oxidations, Reductions, Semiconductor, Solvents, Sonication, Spectra, Surface Science, Transduction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?