EMAIL THIS PAGE TO A FRIEND
U8754 Sigma

Urea tablets

Purchase

Properties

Quality Level   GMP

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
YR6250000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?