• USA Home
  • U9375 - Uricase from Porcine Liver

EMAIL THIS PAGE TO A FRIEND
U9375 Sigma

Uricase from Porcine Liver

Type V, soluble powder, ≥10 units/g protein

Purchase

Properties

type   Type V
form   soluble powder
storage temp.   −20°C

Description

Linkage

This type usually has a somewhat higher activity than our Type II. It is also more soluble and is probably more suitable for the routine determination of uric acid per many of the automated and manual colorimetric procedures. Some automated procedures call for less uricase than is actually needed to completely oxidize the uric acid. We suggest you evaluate the amount required in your own laboratory.

Unit Definition

One unit will convert 1.0 μmole of uric acid to allantoin per min at pH 8.5 at 25°C, unless otherwise indicated.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YU7100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?