• USA Home
  • V0877 - Snake venom from Crotalus basiliscus

EMAIL THIS PAGE TO A FRIEND
V0877 Sigma

Snake venom from Crotalus basiliscus

Synonym: Mexican West-Coast Rattlesnake

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   −20°C

Description

Packaging

100 mg in glass bottle

Application

Snake venom from Crotalus basiliscus (Mexican West-Coast Rattlesnake) may be used as a source of α 2PI (α 2-antiplasmin) degrading and α 2M (α 2-macroglobulin) inhibiting enzymes; fibrinolytic enzyme as well as other toxins. It may also be used as an immunogen or for proteome research.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?