• USA Home
  • V0879 - [pGlu16]-Vasoactive Intestinal Peptide Fragment 16-28 Chicken

EMAIL THIS PAGE TO A FRIEND
V0879 Sigma

[pGlu16]-Vasoactive Intestinal Peptide Fragment 16-28 Chicken

≥97% (HPLC)

Synonym: [pGlu16]-VIP 16-28

  • CAS Number 73073-47-9

  • Empirical Formula (Hill Notation) C67H113N17O18S

  • Molecular Weight 1476.78

  •  MDL number MFCD00133972

  •  PubChem Substance ID 24900700

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   AHQRVCDJIDPDFQ-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Glp-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Val-Leu-Thr-NH2

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?