EMAIL THIS PAGE TO A FRIEND
V119 Sigma

Verruculogen

powder

Synonym: 1,10,10a,14,14a,15b-Hexahydro-10,10a-dihydroxy-7-methoxy-2,2-dimethyl-5-(2-methyl-1-propenyl)-5H,12H-3,4-dioxa-5a,11a,15a-triazacyclooct[1m]indeno[5,6-b]fluorene-11,15(2H,13H)-dione, TR 1

Purchase

Properties

InChI Key   LRXYHMMJJCTUMY-GWXUGYLUSA-N
form   neat
  powder
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   chloroform: 20 mg/mL
format   neat
storage temp.   −20°C

Description

Biochem/physiol Actions

Selective blocker of high conductance Ca2+-activated (Maxi-K) potassium channels.

Caution

Photosensitive

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3
RTECS 
JH4920000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?