• USA Home
  • V1251 - Snake venom from Pseudechis porphyriacus

EMAIL THIS PAGE TO A FRIEND
V1251 Sigma

Snake venom from Pseudechis porphyriacus

Synonym: Common Black

Purchase

Description

Application

Source of phospholipase A2.

Packaging

10, 100 mg in serum bottle

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?