EMAIL THIS PAGE TO A FRIEND
V1390 Sigma

Vapreotide

Synonym: RC 160

  • Empirical Formula (Hill Notation) C57H70N12O9S2

  • Molecular Weight 1131.37

Purchase

Properties

form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Tnfrsf8(25069)

Description

Amino Acid Sequence

DPhe-Cys-Tyr-DTrp-Lys-Val-Cys-Trp-NH2

Biochem/physiol Actions

Somatostatin analogue shown to inhibit tumor growth. Vapreotide lacks important enzyme cleavage sites allowing it to be more stable than somatostatin.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?