• USA Home
  • V1755 - Scorpion venom from Leiurus quinquestriatus hebraeus

EMAIL THIS PAGE TO A FRIEND
V1755 Sigma

Scorpion venom from Leiurus quinquestriatus hebraeus

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Scorpion, Toxins and Venoms
storage temp.   −20°C

Description

Biochem/physiol Actions

Source of chlorotoxin, an intracellular blocker of low-conductance Cl- channels; and charybdotoxin, a blocker of voltage-sensitive Ca2+-activated K+ channels.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX3965400

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?