• USA Home
  • V1888 - Visualization of Receptors in situ: Applications of Radioligand Binding
EMAIL THIS PAGE TO A FRIEND
V1888 Sigma
Visualization of Receptors in situ: Applications of Radioligand Binding
  •  ISBN-10 0-8493-0042-8
  •  ISBN-13 978-0-8493-0042-4
Purchase
Properties
Related Categories Biotechnology Books, Books and Software, Labware More...
publication info   E. Moyse and S.M. Krantic, CRC Press, 2001, 208 pp., hard cover
mfr. no.   CRC Press
Description
General description
Radioligand binding in situ is the most sensitive method for detecting available receptors. But the success and reliability of this approach are determined by a number of technical requirements and constraints. This book combines theory and practice. It covers procedures such as radioligand and tissue preparations, in vitro assays, autoradiographic emulsion handling and image analysis. It also provides detailed protocols and uses parallel columns to convey practical comments and advise, often not encountered in original research papers.
Table of Contents
LIGHT MICROSCOPY. Ligand Labeling with Radioisotopes. Tissue Preparation for Light Microscopy. Radioligand Binding Assay on Frozen Tissue Sections. Film (Dry) Radioautography. Liquid Emulsion (Wet) Radioautography. Quantification of Radioautographic Labeling. ELECTRON MICROSCOPY. Choice of Radioligand. Tissue Preparation. Radioligand Binding Assay on Thick Tissue Slices. Embedding and Ultrathin Sectioning. Radioautography. Coloration and Contrasting of Ultrathin Sections. Analysis and Quantification of Radioautographic Labeling.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?