• USA Home
  • V2132 - [Lys1, Pro2,5, Arg3,4, Tyr6]-Vasoactive Intestinal Peptide human, porcine, rat

EMAIL THIS PAGE TO A FRIEND
V2132 Sigma

[Lys1, Pro2,5, Arg3,4, Tyr6]-Vasoactive Intestinal Peptide human, porcine, rat

≥97% (HPLC)

Synonym: VIP Antagonist, VIPi, [Lys1, Pro2,5, Arg3,4, Tyr6]-VIP

  • CAS Number 125093-93-8

  • Empirical Formula (Hill Notation) C154H257N49O40S

  • Molecular Weight 3467.06

  •  MDL number MFCD00133967

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Gastrointestinal Peptides, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   FBYWUGLFWCEKAN-KQQCXCAZSA-N
assay   ≥97% (HPLC)
composition   Peptide content, ~70%
storage temp.   −20°C
Gene Information   human ... VIP(7432)

Description

Amino Acid Sequence

Lys-Pro-Arg-Arg-Pro-Tyr-Thr-Asp-Asn-Tyr-Thr-Arg-Leu-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Asn-Ser-Ile-Leu-Asn-NH2

Application

[Lys1, Pro2,5, Arg3,4, Tyr6]-Vasoactive Intestinal Peptide human, porcine, rat wasused as a test compound to study the production of cyclic AMP (cAMP) by different polypeptides in cerebral cortical slices of the chick.

Biochem/physiol Actions

VIP receptor antagonist. Blocks VIP neurotransmission in the exocrine pancreas, but does not block the effects of PACAP, which shares 68% homology with VIP. In contrast, it blocks both VIP- and PACAP-stimulated cyclic AMP accumulation in an osteoblast-like cell line.

[Lys1, Pro2,5, Arg3,4, Tyr6]-Vasoactive Intestinal Peptide human, porcine, rat is a neuropeptide synthesized in the central nervous system that enhances the formation of adenosine 3′,5′-cyclic monophosphate in cerebral cortex and hypothalamus of chick and rat. VIP also modulates the cholinergic as well as serotonergic neurotransmitter systems.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?