• USA Home
  • V3125 - Bee venom from Apis mellifera (honey bee)

EMAIL THIS PAGE TO A FRIEND
V3125 Sigma

Bee venom from Apis mellifera (honey bee)

dried venom

Purchase

Properties

Related Categories Bee, Cell Biology, Cell Signaling and Neuroscience, Toxins and Venoms
form   dried venom
storage temp.   −20°C

Description

Other Notes

The entire non-volatile part of the venom

Preparation Note

Obtained by stimulating bees to sting a sheet from which the venom is then gathered and dried.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX3958700

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?