• USA Home
  • V3129 - Snake venom from Oxyuranus scutellatus

EMAIL THIS PAGE TO A FRIEND
V3129 Sigma

Snake venom from Oxyuranus scutellatus

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
suitability   suitable for activation of prothrombin (per Owen, W.G. and Jackson, C.M., Thrombosis Res., 3, 705 (1973).)
storage temp.   −20°C

Description

General description

This venom may be from subspecies canni (Papuan Taipan) or scutellatus (Australian Taipan) or a mixture from both. Physical characteristics are almost identical.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX3994000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?