• USA Home
  • V3376 - Snake venom from Micrurus carinacauda demerilii

EMAIL THIS PAGE TO A FRIEND
V3376 Sigma

Snake venom from Micrurus carinacauda demerilii

Synonym: South American Coral Snake

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?