• USA Home
  • V3508 - Anti-Vasoactive Intestinal Peptide (VIP) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
V3508 Sigma

Anti-Vasoactive Intestinal Peptide (VIP) antibody produced in rabbit

whole antiserum

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurotransmission, Antibodies to Neuropeptides, Neuroscience,
species reactivity   rat
application(s)   dot blot: 1:10,000
  immunohistochemistry (frozen sections): 1:8,000 using 4% paraformaldehyde perfusion-fixed, frozen sections of rat brain
clone   polyclonal
antibody form   whole antiserum
contains   15 mM sodium azide
shipped in   dry ice
storage temp.   −20°C
Gene Information   rat ... Vip(117064)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic vasoactive intestinal peptide (VIP).

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?