• USA Home
  • V3626 - Spider venom from Latrodectus mactans (Black Widow)

EMAIL THIS PAGE TO A FRIEND
V3626 Sigma

Spider venom from Latrodectus mactans (Black Widow)

venom glands

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, S,
form   venom glands
shipped in   dry ice
storage temp.   −20°C

Description

Packaging

Each set consists of a freshly frozen sac containing a pair of venom glands.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?