• USA Home
  • V3631 - Freezing vials, natural
EMAIL THIS PAGE TO A FRIEND
V3631 Sigma
Freezing vials, natural
volume 1,500 μL, style conical, non-sterile
Purchase
Properties
Related Categories Cryo Vials, Cryoware, Labware More...
material   style conical
sterility   non-sterile
mfr. no.   Sigma-Aldrich
volume   1,500 μL
color   natural Cap
Description
General description
Manufactured from an exceptionally clear, virgin polypropylene with a medical-grade silicone O-ring seal. These vials are designed for safe storage at temperatures as low as -196°C but should be used only in the gas phase of liquid nitrogen.
The specially formulated silicone O-ring ensures a positive leakproof seal at all temperatures. A unique base on the skirted vials locks them into place in the microtube locking work station (Z376809). Tubes can be autoclaved to 121°C in upright position with caps loosened. They also can be centrifuged up to 17,000 × g.
Packaging
Caps are bagged separately from tubes
For non-sterile vials, caps and tubes are bagged separately.
For sterile vials, caps and tubes are assembled in bags.
Price and Availability
Cell Freezing
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?