• USA Home
  • V4125 - Snake venom from Agkistrodon contortrix mokason (Northern Copperhead)

EMAIL THIS PAGE TO A FRIEND
V4125 Sigma

Snake venom from Agkistrodon contortrix mokason (Northern Copperhead)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   −20°C

Description

Packaging

100 mg in glass bottle

Application

Snake venom from Agkistrodon contortrix mokason (Northern Copperhead) may be used as a source of disintegrins, fibrin(ogen)olytic activities, phospholipase A2 as well as other toxins. It may also be used as an immunogen or for proteome research.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?