• USA Home
  • V4375 - Snake venom from Agkistrodon piscivorus leukostoma

EMAIL THIS PAGE TO A FRIEND
V4375 Sigma

Snake venom from Agkistrodon piscivorus leukostoma

Synonym: Western Cottonmouth Moccasin

Purchase

Description

Packaging

1 g in glass bottle

25 mg in glass bottle

Price and Availability


DISCOVER Bioactive Small Molecules

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX3970600

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?