• USA Home
  • V4378 - Snake venom from Naja naja

EMAIL THIS PAGE TO A FRIEND
V4378 Sigma

Snake venom from Naja naja

Synonym: Common Indian Cobra

Purchase

Description

Packaging

25 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?