• USA Home
  • V4383 - Vimentin from bovine lens

EMAIL THIS PAGE TO A FRIEND
V4383 Sigma

Vimentin from bovine lens

lyophilized powder

Purchase

Description

Other Notes

A component of mesenchymal cell intermediate filaments.

Reconstitution

When reconstituted with distilled water at 1 mg vimentin per mL, the solution will contain 10 mM sodium phosphate, pH 7.5, 2 mM dithiothreitol, 1 mM EDTA, 6 M urea, and 10 mM methylammonium chloride.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?