• USA Home
  • V5503 - Wasp venom (social) from Polistes apachus

EMAIL THIS PAGE TO A FRIEND
V5503 Sigma

Wasp venom (social) from Polistes apachus

Synonym: (Paper Wasp)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Toxins and Venoms, Wasp
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?