• USA Home
  • V5638 - vivoSIS Cell Culture Products
EMAIL THIS PAGE TO A FRIEND
V5638 Sigma
vivoSIS Cell Culture Products
dried sheets, L × W 75 mm × 25 mm
Purchase
Properties
Related Categories Cell Culture Supplies, Coated Cultureware, Labware More...
L × W   75 mm × 25 mm
Description
General description
vivoSIS¿ Cell Culture Products are extracellular matrix substrates with three-dimensional architecture. The material contains growth factors, proteoglycans, glycoproteins, glycoaminoglycans and at least three types of collagen. The extracellular matrix is not reconstituted, but is derived from a natural source - porcine small intestine submucosa. As shown in the SEM¿s, the architecture of each side is different. The sides also have different porosities and permeabilities. These products have been used for various cell culture applications, including evaluation of chemotaxis or cell adhesion and for use as a substrate for cell differentiation and cell proliferation. They are NOT for use in humans or in vitro diagnostics.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?