• USA Home
  • V5753 - Wasp venom (social) from Dolichovespula arenaria (Yellow Hornet)

EMAIL THIS PAGE TO A FRIEND
V5753 Sigma

Wasp venom (social) from Dolichovespula arenaria (Yellow Hornet)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Toxins and Venoms, Wasp
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?