• USA Home
  • V6257 - Monoclonal Anti-Vasoactive Intestinal Peptide Receptor 2 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
V6257 Sigma

Monoclonal Anti-Vasoactive Intestinal Peptide Receptor 2 antibody produced in mouse

1.0 mg/mL, clone AS69, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-VIPR 2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Neurotransmission, Antibodies to Neurotransmitter Receptors, Neuroscience,
species reactivity   rat, human
application(s)   immunohistochemistry (frozen sections): 0.25-2 μg/mL using human VIPR 2 expressed on rat hippocampal cells fixed with paraformaldehyde followed by 0.1% Triton X-100
  western blot: 0.5-2 μg/mL
clone   AS69, monoclonal
concentration   1.0 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt 49 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... VIPR2(7434)
rat ... Vipr2(29555)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

unique peptide corresponding to a portion of human vasoactive intestinal peptide receptor 2.

Physical form

Solution in 0.2 μm sterile-filtered phosphate buffered saline with 0.08% sodium azide.

Preparation Note

Purified from tissue culture supernatant.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

WGK Germany  -
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?