• USA Home
  • V7878 - Val-Pro hydrochloride

EMAIL THIS PAGE TO A FRIEND
V7878 Sigma

Val-Pro hydrochloride

  • CAS Number 105931-64-4

  • Empirical Formula (Hill Notation) C10H18N2O3 · HCl

  • Molecular Weight 250.72

  •  MDL number MFCD00191045

  •  PubChem Substance ID 24900768

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   IIQAHNBGJJXSGP-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?