EMAIL THIS PAGE TO A FRIEND
V7887 Sigma

VSV-G Peptide

≥97% (HPLC), lyophilized powder

Purchase

Properties

Related Categories Molecular Biology, Proteomics, Purification and Detection, Recombinant Protein Expression and Analysis, VSV-G More...
assay   ≥97% (HPLC)
form   lyophilized powder
mol wt   mol wt 1339.5 Da
storage temp.   −20°C

Description

Amino Acid Sequence

Tyr-Thr-Asp-Ile-Glu-Met-Asn-Arg-Leu-Gly-Lys

Other Notes

Lyophilized from 0.1% TFA in H2O

Price and Availability


Amplified Detection

Amicon Pro Purification System
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?