• USA Home
  • V8130 - Freezing vials 2.0 mL, sterile
EMAIL THIS PAGE TO A FRIEND
V8130 Sigma
Freezing vials 2.0 mL, sterile
red cap
Purchase
Properties
Related Categories Cryo Vials, Cryoware, Labware More...
sterility   sterile; γ-irradiated (Certified RNase-, DNase-, Pyrogen- and DNA-free.)
color   red cap
Description
General description
Graduated with marking square.
Color coded caps have internal threads and O-ring seal to minimize contamination.
May be autoclaved with loose caps.
Do not immerse in the liquid phase of liquid nitrogen.
Price and Availability
Cell Freezing
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?