EMAIL THIS PAGE TO A FRIEND
V8134 Sigma
Nunc® CryoTubes®
cryogenic vial, 3.6 mL, internal thread, round bottom, starfoot, freestanding, 1600/cs
Synonym: cryogenic tubes, cryogenic vials
Purchase
Properties
Related Categories Cryo Vials, Cryotube Vials, Cryoware, Labware, Manufacturer Browser,
material   cap screw top
  polypropylene
  polypropylene
  tube round bottom
  natural tube
sterility   sterile (with writing area)
feature   internal thread; screw cap
packaging   case of 1600
mfr. no.   Nunc, 379189
L × diam.   72 mm × 12.5 mm
working volume   3.6 mL
Description
General description
Nunc® CryoTubes®, Internal Thread
• Intended for cryogenic transportation and storage of biological material
• CE marked for IVD
• Conform to the IATA requirements for the transport of diagnostic specimens
• Perform well in mechanical freezers, and in nitrogen (gas and liquid* phase)
• One hand operation with starfoot tubes in CryoTube® Rack (R1152; R1277)
• Non-toxic, USP Class VI
• Non-pyrogenic
• Non-mutagenic
• Sterile (SAL 10-6) according to ISO 11137
• Re-closable zippered bags
• Internal thread with a silicone gasket provides the best possible seal
• Catalog No. and Lot No. are printed on the bags
*For extreme storage conditions like liquid phase of nitrogen CryoFlex Tubing must be used.
Legal Information
CryoFlex is a trademark of Thermo Fisher Scientific or its subsidiaries
CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Cell Freezing
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?