• USA Home
  • V8254 - Vindesine sulfate salt

EMAIL THIS PAGE TO A FRIEND
V8254 Sigma

Vindesine sulfate salt

≥95% (TLC)

Synonym: Desacetylvinblastine amide sulfate salt

Purchase

Properties

assay   ≥95% (TLC)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Binds to tubulin and arrests the cell cycle in metaphase by preventing tubulin polymerization and depolymerizing previously formed microtubule.

Other Notes

Semisynthetic derivative of the antineoplastic vinblastine.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
YY8090000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?