• USA Home
  • V8504 - Multiwell plate washer/dispenser manifolds
EMAIL THIS PAGE TO A FRIEND
V8504 Sigma
Multiwell plate washer/dispenser manifolds
3-way valve for repeat kit
Purchase
Description
General description
For use with 96 well multiwell plates
• Dispense or aspirate media, or wash, a full row or column at a time
• Luer fitting
• Stainless steel and PTFE construction for easy cleaning and autoclavability
• Optional repeat kit provides continuous operation capability
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?