• USA Home
  • V9625 - Snake venom from Naja nivea

EMAIL THIS PAGE TO A FRIEND
V9625 Sigma

Snake venom from Naja nivea

Synonym: Naja flava; Cape Cobra

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Snake, Toxins and Venoms
storage temp.   −20°C

Description

Packaging

100 mg in poly bottle

Application

Snake venom from Naja nivea (cape cobra) may be used as a source of protease inhibitors as well as other toxins. It may also be used as an immunogen or for proteome research.Source of postsynaptic neurotoxins

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YX3992800

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?