• USA Home
  • W0374 - Workshields, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
W0374 Sigma
Workshields, acrylic, for β-emitters
size 14 in. × 21 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
size   14 in. × 21 in.
thickness   3/8 in.
Description
General description
Base is molded with a special bend for use with Spill Trays, q.v.
Free-standing shields of clear acrylic provide effective protection against most β-emitting isotopes, including 32P. Angled fronts permit easy viewing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?