• USA Home
  • W1377 - Workshields, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
W1377 Sigma
Workshields, acrylic, for β-emitters
base size 12 in. × 12 in., H 18 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
description   with side shields
H   18 in.
base size   12 in. × 12 in.
thickness   1/2 in.
Description
General description
Free-standing shields of clear acrylic provide effective protection against most β-emitting isotopes, including 32P. Angled fronts permit easy viewing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
Z677809 Acrylic workshield for ß-emitters, Medium, L × W × H 305 mm × 356 mm × 527 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?