• USA Home
  • W1874 - Workshields, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
W1874 Sigma
Workshields, acrylic, for β-emitters
size 22 in. × 16 in. × 20 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
size   22 in. × 16 in. × 20 in.
Description
General description
Constructed of 3/8 in. acrylic, provides the best all-around protection while working with β-emitting radioisotopes. Closed sides and back also protect others nearby. Vertical sliding doors open wide for placing and removing materials, and close down to small openings for two-hand access during work. Built-in pipet rack holds two pipettes. Small ports in back for electrical, vacuum, and exhaust access. Designed to fit inside 21 x 27 in. spill tray for complete protection and easy cleanup of spills.
Free-standing shields of clear acrylic provide effective protection against most β-emitting isotopes, including 32P. Angled fronts permit easy viewing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?