• USA Home
  • W2001 - Workshields, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
W2001 Sigma
Workshields, acrylic, for β-emitters
base size 12 in. × 17 in., H 20 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
H   20 in.
base size   12 in. × 17 in.
thickness   3/8 in.
Description
General description
Free-standing shields of clear acrylic provide effective protection against most β-emitting isotopes, including 32P. Angled fronts permit easy viewing.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z677817 Acrylic workshield for ß-emitters, Large, L × W × H 314 mm × 406 mm × 521 mm
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?