EMAIL THIS PAGE TO A FRIEND
W4136 Sigma
PCR Workstation
W × D × H 30 in. × 24 in. × 24 in., AC input 240 V, Euro plug
Purchase
Properties
Related Categories Labware, Molecular Biology Supplies, PCR (Polymerase Chain Reaction) Supplies, PCR Workstation
W × D × H   30 in. × 24 in. × 24 in.
Description
General description
A self-contained work area that will help protect your PCR runs against contamination. Dual UV bulbs irradiate the work area prior to use, reducing the possibility that contaminating DNA will be amplified. Containment features reduce the chance of airborne contamination.
The non-ventilated hood has acrylic windows for a clear view, an acid-resistant black formica work surface, a laminated safety glass fascia, and double access doors. The germicidal lamps are controlled by a 24-hr timer. The work light is provided by dual fluorescent lamps, and there is a ceiling-mounted double AC outlet. Replacement part sold separately.
Legal Information
PCR Workstation is a trademark of C.B.S. Scientific Co.
Price and Availability
Your Key to Success
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?