• USA Home
  • W4503 - Portable work station, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
W4503 Sigma
Portable work station, acrylic, for β-emitters
L × W × H 12 3/4 in. (32 cm) × 15 in. (38 cm) × 3 7/8 in. (10 cm), thickness 3/8 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Workshields, Safety Equipment
material   acrylic lid and box
L × W × H   12 3/4 in. (32 cm) × 15 in. (38 cm) × 3 7/8 in. (10 cm)
thickness   3/8 in.
Description
General description
With the lid open toward the user, the unit is a shielded work station. When closed, it is a box for safe storage and transport of β-emitters.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?