EMAIL THIS PAGE TO A FRIEND
W4512 Sigma

Well orienter

48 wells

Purchase

Properties

Related Categories Labware, Multiwell Plates, Washers, Orienters, etc. More...
sterility   non-sterile
color   none
matrix active group   Non-treated surface treatment

Description

General description

A perfectly sized plastic card with sharp, alpha-numeric characters to identify each and every well on a microplate, will end cross referencing headaches. Once positioned under the microplate, the Well Orienter makes identifying sample wells easy and headache-free!

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?