• USA Home
  • WGHT - Wright-Giemsa Stain Set for Hema-Tek® 1, 1000 and 2000

EMAIL THIS PAGE TO A FRIEND
WGHT Sigma

Wright-Giemsa Stain Set for Hema-Tek® 1, 1000 and 2000

  •  eCl@ss 42039003

Purchase

Properties

Related Categories Cell Biology, Hematology Stains, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Application

Set includes a single reagent Wright Stain with two rinse solutions.

General description

Specific packaging for Hematek automatic stainers.

Legal Information

Hema-Tek is a registered trademark of Siemens Healthcare Diagnostics, Inc.

Price and Availability


All labs need water

Standard component

Description

Product #

Add to Cart

Rinse Solution 1 300 mL SDS    
Rinse Solution 2 900 mL SDS    
Wright-Giemsa Stain (0.15% in methanol with stabilizers) 300 mL SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?