Synonym: Anti-PARK8, Anti-ROCO2, Anti-leucine-rich repeat kinase 2
| Related Categories | 3-3B, Alphabetical Index, Antibodies, Antibodies for Parkinson’s Disease Research, Cell Biology, |
| species reactivity | human |
| application(s) | indirect ELISA: suitable |
| western blot: suitable | |
| clone | 3B2, monoclonal |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| isotype | IgG1κ |
| immunogen sequence |
NSRNASIWLGCGHTDRGQLSFLDLN |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... LRRK2(120892) |
LRRK2 (AAV63975, a.a. 2161-2260) partial recombinant protein with GST tag. MW of the GST tag alone is 26 kDa.
Solution in phosphate buffered saline, pH 7.2
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?