• USA Home
  • WSGD128 - Wright Stain for Geometric Data Hemastainer and Hematrak

EMAIL THIS PAGE TO A FRIEND
WSGD128 Sigma

Wright Stain for Geometric Data Hemastainer and Hematrak

Purchase

Properties

Related Categories Cell Biology, Hematology Stains, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Application

Intended for batch staining on the Geometric Data Hemastainer, or similar dip and dunk style stainers.

Other Notes

Wright stain, modified, 0.14%, with stabilizers in methanol.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?