• USA Home
  • WSHTS - Wright Stain sets for Hema-Tek® I

EMAIL THIS PAGE TO A FRIEND
WSHTS Sigma

Wright Stain sets for Hema-Tek® I

Purchase

Properties

Related Categories Cell Biology, Hematology Stains, Hematology and Histology More...
Quality Level   GMP-IVD
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

General description

Specific packaging for Hema-tek automatic stainer.

Packaging

Set includes a single-reagent Wright stain with two rinse solutions.

Legal Information

Hema-Tek is a registered trademark of Siemens Healthcare Diagnostics, Inc.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3316 9
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?