• USA Home
  • X0502 - Xylan from birch wood

EMAIL THIS PAGE TO A FRIEND
X0502 Sigma

Xylan from birch wood

Xylose residues ≥90 % by HPAE

Synonym: Poly(β-D-xylopyranose[1→4])

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates P-Z, Polysaccharide More...
composition   Xylose residues, ≥90% HPAE

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?